Xylene Substitute, Electron Microscopy Sciences
Product Details & Documents
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
- Minimal tissue shrinkage
- Dries slowly with no residue
- Less toxic than xylene
- Soluble with alcohol, paraffin embedding media, and mounting media
- Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.
Specifications
Specifications
- Pack type:Bottle
- Description:Xylene substitute
- Size:1 gal.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.