Orders LinkContactUsLinkComponent
Count Up/Count Down Timer, Electron Microscopy Sciences
Count Up/Count Down Timer, Electron Microscopy Sciences
Timer, count up count down
  100502-098
Supplier #:  72603
Count Up/Count Down Timer, Electron Microscopy Sciences
  100502-098
Count Up/Count Down Timer, Electron Microscopy Sciences
Count Up/Count Down Timer, Electron Microscopy Sciences
Timer, count up count down
$79.37
Each
Count up or down for 100 hours in intervals of 1 second.
  • Alarm clock mode
  • Its memory allows for the repeat of a cycle just completed
In the count down mode its unique feature tells how much time has elapsed after the alarm sounds.
Dimensions: 2³/₄×2³/₄×¹/₂"
Weight: 1.8 oz.
Certifications: With N.I.S.T. traceable certificate of compliance.
Delivery information: Comes complete with a built in stand, pocket clip, button cell battery, and magnet.
  • Description:
    Timer, count up count down
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.