Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Micro-Tool Individual Micro Tools, Electron Microscopy Sciences

Micro-Knife 45°, 0.50 mm, Tip 9
Catalog # 100501-642
 :  62091-09-50
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Catalog # 100501-642
Supplier:  
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences
Micro-Tool Individual Micro Tools, Electron Microscopy Sciences

Micro-Knife 45°, 0.50 mm, Tip 9

Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Micro-Tools are the smallest known precision tools available for the laboratory and micro electronics industry. These tools are designed for anyone who is engaged in microscope work, miniature applications as well as specimen manipulation.

  • Consists of a threaded handle (MT1), 4.75" long and an interchangeable threaded base with tool tip
  • Available in 32 different tip configurations with interchangeable handles
  • Tip material is hardened tool steel unless noted
Specifications
  • Description:
    Micro-Knife 45°
  • Size:
    0.50 mm
  • Tip configuration:
    Tip 9

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.