Orders LinkContactUsLinkComponent
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
Horizontal Opening, Straight
Catalog # 100501-486
Supplier #:  77930-01
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
Catalog # 100501-486
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
Horizontal Opening, Straight
Price
$959.14
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Scissors are made from 20 gauge stainless steel and have a 3 mm cutting edge.
Specifications
  • Description:
    Horizontal Opening
  • Curvature:
    Straight
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.