Order Entry
Canada
Orders LinkContactUsLinkComponent
Formalin 10%, TissuePrep, buffered

Formalin 10%, TissuePrep, buffered

60 ml
Catalog # 100500-248
Health
Irritant
Supplier #:  15742-60
Formalin 10%, TissuePrep, buffered
Catalog # 100500-248
CAS Number:  50-00-0
Formalin 10%, TissuePrep, buffered

60 ml

CAS Number:  50-00-0
Price
$164.89
Log in for availability
Case of 20 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ready-to-use, prefilled with 10% formalin.

Prefilled with 10% formalin Cat# 15740. Ready-to-use in pre-filled cups. Labeled in accordance to OSHA requirements, which gives you immediate information regarding handling and first aid procedures. Pre-measured in wide-mouth, leak-proof, opaque polypropylene containers which limits your exposure to formaldehyde and saves you time.

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
Specifications

Specifications

  • Size:
    60 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.