Orders LinkContactUsLinkComponent
Weibel 3 Graticles, Electron Microscopy Sciences
Weibel 3 Graticles, Electron Microscopy Sciences
NGW3 Graticule, 21 mm
Catalog # 100499-228
Supplier #:  68023-21
Weibel 3 Graticles, Electron Microscopy Sciences
Catalog # 100499-228
Weibel 3 Graticles, Electron Microscopy Sciences
Weibel 3 Graticles, Electron Microscopy Sciences
NGW3 Graticule, 21 mm
Price
$774.46
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mertz for stereology. Surface chrome image.
Reference: E.R. Weibel, G.S. Kishtler & W.F. Scherle. 1986. J. Cell Biology.30, 23.
Specifications
  • Description:
    NGW3 Graticule
  • Size:
    21 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)