Orders LinkContactUsLinkComponent
Brushes, Red Sable, Electron Microscopy Sciences
Brushes, Red Sable, Electron Microscopy Sciences
Red Sable Brush, Sable hair, #3
Catalog # 100498-904
Supplier #:  66100-03
Brushes, Red Sable, Electron Microscopy Sciences
Catalog # 100498-904
Brushes, Red Sable, Electron Microscopy Sciences
Brushes, Red Sable, Electron Microscopy Sciences
Red Sable Brush, Sable hair, #3
Price
$28.56
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Specifications
  • Bristle material:
    Sable hair
  • Description:
    Red Sable Brush
  • Size:
    #3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)