Orders LinkContactUsLinkComponent
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Manganese Single
  100495-080
 :  80074-MN
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
  100495-080
 :  
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Manganese Single
 :  
$1,044.76
Each
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.
  • Description:
    Manganese Single
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)