Orders LinkContactUsLinkComponent
Iris Tissue Forceps, Electron Microscopy Sciences | Avantor
Iris Tissue Forceps, Electron Microscopy Sciences
Iris 1x2 Teeth Tissue Forceps, Straight
Catalog # 100494-912
Fournisseur #:  78611-4S
Iris Tissue Forceps, Electron Microscopy Sciences
Catalog # 100494-912
Iris Tissue Forceps, Electron Microscopy Sciences
Iris Tissue Forceps, Electron Microscopy Sciences
Iris 1x2 Teeth Tissue Forceps, Straight
Price
$21.67
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Extra delicate tissue forceps with 1x2 teeth. Made from stainless steel and 4" long. The tips are 1.2 mm wide.
Spécifications
  • Description:
    Iris 1x2 Teeth Tissue Forceps, Straight
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.