Magnabrite™ Light-Gathering Magnifiers, Electron Microscopy Sciences
Magnabrite™ magnifies the viewing area by 4X, while intensifying available light and focusing it on the subject area
Made from solid grounded acrylic (not molded) and polished like a glass lens. Crystal clear and pre-focused. There are no bulbs or batteries needed. Available light is nearly four times brighter in the subject area. Available with 90, 64, or 51 mm diameter base.
An "Infinite eyepoint" eliminates the need to keep the eye close to the magnifier. It can be viewed from any distance, at any angle which reduces eye strain and allows several people to view the same object at once. Base is concave which reduces the risk of scratching the film. The light comes with a drawstring storage pouch.
- Magnification:4×
- Description:90 mm Diameter
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.