Orders LinkContactUsLinkComponent
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton-Tipped Applicator, Non sterile, 1000/Pack
Catalog # 100493-712
Supplier #:  72310-S
Cotton Tipped Applicators, Electron Microscopy Sciences
Catalog # 100493-712
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton-Tipped Applicator, Non sterile, 1000/Pack
Price
$399.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.
Specifications
  • Sterility:
    Non sterile
  • Description:
    Cotton-Tipped Applicator
  • Packaging:
    1000/Pack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)