Orders LinkContactUsLinkComponent
Large Glass Slide Storage Box, Electron Microscopy Sciences
Large Glass Slide Storage Box, Electron Microscopy Sciences
25 Microscope Slide Box, 3x2" Slides
Catalog # 100493-362
Supplier #:  71481-01
Large Glass Slide Storage Box, Electron Microscopy Sciences
Catalog # 100493-362
Large Glass Slide Storage Box, Electron Microscopy Sciences
Large Glass Slide Storage Box, Electron Microscopy Sciences
25 Microscope Slide Box, 3x2" Slides
Price
$148.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
100-Slide Capacity:
These storage boxes are available in two sizes.
100-Slide Capacity:
Durable, wood frame, covered with heavy black embossed paper. The hinges and latches are Nickel plated. A numbered plastic insert provides spacing for the handling of the slides. The box accommodates 100 of 3x2" slides. The box measures 24.7 x 19.7 x 5.7 cm (9 3/4 x 7 4/5 x 2 1/4").
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
Specifications
  • Description:
    25 Microscope Slide Box, 3x2" Slides
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.