Orders LinkContactUsLinkComponent
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Borosilicate, 4"L
Catalog # 100490-958
Supplier #:  70950-15
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Catalog # 100490-958
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Borosilicate, 4"L
Price
$168.41
Log in for availability
Each Pack of 250
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
Specifications
  • Description:
    Borosilicate
  • Length:
    4"L
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)