Orders LinkContactUsLinkComponent
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
1 well, 520 µl, 20 mm dia, 1.7 mm
Catalog # 100490-802
Supplier #:  70327-20
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Catalog # 100490-802
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
1 well, 520 µl, 20 mm dia, 1.7 mm
Price
$284.30
Log in for availability
Each Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Specifications
  • Depth:
    1.7 mm
  • Description:
    1 well, 520 µl
  • Size:
    20 mm dia
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)