Orders LinkContactUsLinkComponent
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
1 well, 300 µl, 20 mm dia, 0.9 mm
Catalog # 100490-800
 :  70327-10
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
Catalog # 100490-800
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
CoverWell™ Imaging Chambers, Electron Microscopy Sciences
1 well, 300 µl, 20 mm dia, 0.9 mm
Price
$477.95
Log in for availability
Each Pack of 40
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Specifications
  • Depth:
    0.9 mm
  • Description:
    1 well, 300 µl
  • Size:
    20 mm dia
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.