Orders LinkContactUsLinkComponent
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
15 Dram (55.5 ml), 64 mm, 32 mm
Catalog # 100490-536
Supplier #:  64248-35
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Catalog # 100490-536
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
15 Dram (55.5 ml), 64 mm, 32 mm
Price
$42.82
Log in for availability
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Specifications
  • Ø int.:
    32 mm
  • Capacity:
    15 Dram (55.5 ml)
  • Height:
    64 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)