Orders LinkContactUsLinkComponent
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Only
Catalog # 100489-830
Supplier #:  70946
Perfect Loop, Electron Microscopy Sciences
Catalog # 100489-830
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Only
Price
$1,355.35
Log in for availability
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
Specifications
  • Description:
    Perfect Loop Only
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)