Orders LinkContactUsLinkComponent
Dispensing Bottles, Electron Microscopy Sciences
Dispensing Bottles, Electron Microscopy Sciences
60 ml (2 oz.)
Catalog # 100489-770
Supplier #:  64292
Dispensing Bottles, Electron Microscopy Sciences
Catalog # 100489-770
Dispensing Bottles, Electron Microscopy Sciences
Dispensing Bottles, Electron Microscopy Sciences
60 ml (2 oz.)
Price
$66.75
Log in for availability
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Low density polyethylene bottles.
  • Tip has a 0.76 mm (0.003") opening
By squeezing the bottle a steady stream of liquid is produced.
Specifications
  • Capacity:
    60 ml (2 oz.)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)