Lab-Tek® II Chamber Slide™ System, Electron Microscopy Sciences
Lab-Tek® II Chamber Slide™ products offer optimal surface for excellent attachment and growth of cells, easily distinguished by the blue mark
- Sterile, Chamber Slide™ products have removable, polystyrene media chamber of 1, 2, 4, and 8-well configuration
- Low fluorescent glass microscope slide (25 x 75 x 1.2 mm) with rounded corners
- Biocompatible adhesive
- Polystyrene cover
- Inert hydrophobic well border printed on slide
- SuperFrost printed area
Lab-Tek® II CC2™ Chamber Slide™ products are the newest addition to our Lab-Tek® II Chamber Slide™ family. CC2 shares several similarities with the original Lab-Tek® II Chamber Slide, however, it’s pronounced difference is that the glass growth surface has been chemically modified. This modification provides an optimal surface for growing fastidious cells such as primary neurons or poorly adherent cell lines. CC2 also are non-auto fluorescent making them ideal for fluorescent applications such as FISH (Fluorescent in situ Hybridization). The growth surface remains stable without refrigeration.
Lab-Tek® II and Lab-Tek II CC2™ Chamber Slide™ Systems all come packed in a sterile sealed tray. 8 units per tray, 2 trays per pack and 12 trays per case.
This design allows for the removal of the media chamber and adhesive from the slide, using a separation device. The blue hydrophobic border is raised slightly and isolates the individual wells, and prevents cross contamination. The slide can be coversliped.
- Description:CC2™ Slide
- No. of chambers:4
- No. of trays:6
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.