Orders LinkContactUsLinkComponent
Disposable Pipettes – Sterile, Electron Microscopy Sciences
1 ml, Bulk
Catalog # 101413-248
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Catalog # 101413-248
Supplier:  
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Disposable Pipettes – Sterile, Electron Microscopy Sciences
1 ml, Bulk
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PIPETTE, STR DISP 1ML PK100 Catalog #101413-236
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Specifications
  • Volume:
    1 ml
  • Packed:
    Bulk
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.