Orders LinkContactUsLinkComponent
Serum Bottles, Electron Microscopy Sciences
2 ml, Serum Bottles, Clear
Catalog # 101412-388
 :  60306-02
Serum Bottles, Electron Microscopy Sciences
Catalog # 101412-388
Serum Bottles, Electron Microscopy Sciences
Serum Bottles, Electron Microscopy Sciences
2 ml, Serum Bottles, Clear
Price
$664.26
Log in for availability
Case of 288 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
 : Meets USP Type I standards.
Specifications
  • Capacity:
    2 ml
  • Color:
    Clear
  • Description:
    Serum Bottles
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.