Orders LinkContactUsLinkComponent
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Catalog # 100500-900
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Catalog # 100500-900
Supplier:  
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CRYOVIAL PP NS 2.0ML BK CS1000 Catalog #66008-748
Product 66008-748 is similar to 100500-900 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
  • Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)