Orders LinkContactUsLinkComponent
Peel Away Disposable Embedding Molds, Electron Microscopy Sciences
Embedding Mold Square Shape, 22 x 22 mm
Catalog # 100500-224
 :  70182
Peel Away Disposable Embedding Molds, Electron Microscopy Sciences
Catalog # 100500-224
Peel Away Disposable Embedding Molds, Electron Microscopy Sciences
Peel Away Disposable Embedding Molds, Electron Microscopy Sciences
Embedding Mold Square Shape, 22 x 22 mm
Price
$838.30
Log in for availability
Case of 288 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For plastic or paraffin embedding; no block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Specifications
  • Description:
    Embedding Mold Square Shape
  • Size:
    22 x 22 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.