Orders LinkContactUsLinkComponent
Concentric Circles Graticle, Electron Microscopy Sciences
Concentric Circles Graticle, Electron Microscopy Sciences
Graticle NE43, 21 mm, Chrome image
Catalog # 100499-084
Supplier #:  67997-21
Concentric Circles Graticle, Electron Microscopy Sciences
Catalog # 100499-084
Concentric Circles Graticle, Electron Microscopy Sciences
Concentric Circles Graticle, Electron Microscopy Sciences
Graticle NE43, 21 mm, Chrome image
Price
$377.80
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For calibrated measurement in a light microscope.
  • Scales in mm and inches available
  • Surface chrome image
  • Extended size range
The microscope graticle can be used for two-way measurement when calibrated as a micrometer.
Specifications
  • Surface:
    Chrome image
  • Description:
    Graticle NE43
  • Size:
    21 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)