Top-Loading Dispensing Bins, Electron Microscopy Sciences
Product Details & Documents
Bins are made from ¹/₄" thick PVC. They are chemical resistant and easy to clean.
- Removable top for easy loading
- Hinged clear acrylic door
- Different sizes for your needs
Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Specifications
- Description:Small Top-Loading Dispensing Bins
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)