Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
Product 100501-436 is similar to 100496-328 but manufacturer and specifications may vary.
Product 100501-432 is similar to 100496-328 but manufacturer and specifications may vary.
Product 100501-438 is similar to 100496-328 but manufacturer and specifications may vary.
Product Details & Documents
Made from durable polystyrene (with heavy walls).
This material is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25 x 75 mm microscope slides. available in 5 color coded boxes making the boxes easy to identify. Each box measures 222 x 172 x 35 mm (8.75 x 6.75 x 3.375").
Documents
EMS-Core Sampling Tools
Product Literature
Specifications
- Description:100 Capacity
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)