Orders LinkContactUsLinkComponent
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Vannas Scissors Angle On Side, Delicate, 3 mm (⅛"), Extra fine
Catalog # 100493-886
Supplier #:  72932-04
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Catalog # 100493-886
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Vannas Scissors Angle On Side, Delicate, 3 mm (⅛"), Extra fine
Price
$851.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Vannas micro scissors have extra fine tips and are 3" (75 mm) long. Available with a 3 mm or 5 mm cutting edge.
Specifications
  • Description:
    Vannas Scissors Angle On Side, Delicate
  • Blade length:
    3 mm (⅛")
  • Blade type:
    Extra fine
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)