Order Entry
Canada
Orders LinkContactUsLinkComponent
Electrodes for Checker® pH Tester, Electron Microscopy Sciences | Avantor

Electrodes for Checker® pH Tester, Electron Microscopy Sciences

HI 1271 replacement electrode, Model 70699-02, Screw type, 0 - 14
  100493-012
Fournisseur #:  70700-01
Electrodes for Checker® pH Tester, Electron Microscopy Sciences
  100493-012
Electrodes for Checker® pH Tester, Electron Microscopy Sciences
Electrodes for Checker® pH Tester, Electron Microscopy Sciences

HI 1271 replacement electrode, Model 70699-02, Screw type, 0 - 14

 
$57.34
   
Each
 
   
 

Replacement electrodes for Checker® tester.

The extended family of Checker® pH testers now includes standard gel-filled electrodes with built-in BNC or screw type connectors.

 
  • Description:
    HI 1271 replacement electrode
  • For use with:
    Model 70699-02
  • pH range:
    0 - 14
  • Connector:
    Screw type

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.