Scintillation Vial Rack, Electron Microscopy Sciences
Product Details & Documents
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
Specifications
- Description:Scintillation vial rack with handle
- No. of places:24
- W×D×H:11¾×5×3¾"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)