Perfect Loop, Electron Microscopy Sciences
Product Details & Documents
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
Specifications
- Description:Light Microscopy Perfect Loop Set
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)