Orders LinkContactUsLinkComponent
Disposable Glass Tubes, Electron Microscopy Sciences
Yellow Label, 100 mm, 13 mm
Catalog # 100490-622
 :  64636-Y
Disposable Glass Tubes, Electron Microscopy Sciences
Catalog # 100490-622
Disposable Glass Tubes, Electron Microscopy Sciences
Disposable Glass Tubes, Electron Microscopy Sciences
Yellow Label, 100 mm, 13 mm
Price
$629.63
Log in for availability
Each Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tubes are made from borosilicate glass, round bottom and plain top.
The tubes measuring 16 mm in outer diameter and 125 mm in length are Flint glass only.
Labeling area in five pastel colors. Handy fused-in color patches, accept any type of ink or pencil.
Specifications
  • Ø:
    13 mm
  • Description:
    Yellow Label
  • Height:
    100 mm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)