PTFE Printed Slides, Electron Microscopy Sciences
Product Details & Documents
These slides are extremely hydrophobic, acetone proof, chemical resistant and autoclavable. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests.
- Super hydrophobic mask
- Clean, wettable wells
- A bioadhesive surface treatment to enhance cell and tissue attachment
The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities within the well. These slides minimize the use of expensive purified antigens and antibodies. The slides measure 3 x 1" (76 x 26 x 1 mm).
Manufactured to order offering maximum shelf life. The shelf life of 18 months to two years. However, it is recommended that all glass slides (Printed or Un-printed) be used in a timely manner to avoid glass "weathering", (slides becoming cloudy or hazy even with proper storage. All printed slides are autoclavable and have a frosted end. They are intended for one time use only.
Specifications
- Description:Hydrophobic printed slides, 3 well
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)