Orders LinkContactUsLinkComponent
Glassine Envelopes, Electron Microscopy Sciences
Glassine Envelopes, Electron Microscopy Sciences
Plain, 4x5"
Catalog # 100488-560
Supplier #:  74064
Glassine Envelopes, Electron Microscopy Sciences
Catalog # 100488-560
Glassine Envelopes, Electron Microscopy Sciences
Glassine Envelopes, Electron Microscopy Sciences
Plain, 4x5"
Price
572,08 $
Log in for availability
Each Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Glassine envelopes, acid free for negative storage. Open end, thumb-cut, side-seamed.
Specifications
  • Description:
    Plain
  • Size:
    4x5"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)