Serum Bottles, Electron Microscopy Sciences
Product Details & Documents
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
Specifications
- Capacity:10 ml
- Color:Clear
- Description:Serum Bottles
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)