Orders LinkContactUsLinkComponent
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Weighted Beaker/Flask Holder, 250 mL
Catalog # 100498-820
Supplier #:  64441-02
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Catalog # 100498-820
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Weighted Beaker/Flask Holder, Electron Microscopy Sciences
Weighted Beaker/Flask Holder, 250 mL
Price
$464.51
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This weighted holder grips the beaker or flask firmly in four flared stainless steel prongs
The Vikem® vinyl-coated lead ring secured beneath the beaker keeps it immersed in water, providing a low center of gravity which allows water to circulate underneath the beaker.
Specifications
  • Description:
    Weighted Beaker/Flask Holder, 250 mL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.