Sample Storage Sets; Snap Cap Shell Vial, Electron Microscopy Sciences
Product Details & Documents
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom.
Alpha-numerical indexing system allows quick identification and location of stored items.
Ordering information: For vial sizes of 10 and 12 dram please call us for prices and availability.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Specifications
- Cap included:Yes
- Cap type:Snap cap
- Capacity:10.40 ml
- Description:Shell vial series, dram 3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)