Orders LinkContactUsLinkComponent
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Block of 20 Standards X-Ray
Catalog # 100495-014
 :  80070-20
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Catalog # 100495-014
Supplier:  
CAS Number:  
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Customized Multielement X-ray Microanalysis Standards, Electron Microscopy Sciences
Block of 20 Standards X-Ray
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
  • All standards are polished to a 0.25 micron diamond finish and carbon coated
  • They are supplied with fully authenticated certificates of analyses and a location map for standard identification
Pure metals and compounds mounted in brass blocks. Carousel configuration is supplied at an extra cost of (52.00) to the above prices. Please note that for this configuration there is a maximum of 25 standards permitted.
A Faraday Cup, for accurate specimen current measurements, is available as an optional extra on all mounts. The standards are available in different sizes of brass holders, including those of 25 or 32 mm diameter, containing up to 60 standards of your choice. Carousels, 13 mm diameter blocks (to fit pin stubs), and single standards of 5 mm, 3 mm, 2 mm diameter, are also available. Specials can be "tailor-made" to suit individual needs and specific stage configurations.
Specifications
Specifications
  • Description:
    Block of 20 Standards X-Ray
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.