Orders LinkContactUsLinkComponent
Histology Containers, Electron Microscopy Sciences
Histology Containers, Electron Microscopy Sciences
Catalog # 100490-450
Histology Containers, Electron Microscopy Sciences
Catalog # 100490-450
Supplier:  
Histology Containers, Electron Microscopy Sciences
Histology Containers, Electron Microscopy Sciences
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
HISTOLOGY CONTAINER, 250ML Catalog #10153-758
Product 10153-758 is similar to 100490-450 but may have a different manufacturer and/or packaging.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing
  • Designed for storage and transport of histology specimens
  • Made from durable polypropylene (except 8L size which is made from polyethylene).
  • Leak proof seal caps.
The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.
All of the containers (except the 5 mL size) are graduated on three sides.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)