Orders LinkContactUsLinkComponent
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
3.00 ml, Yes
Catalog # 100489-536
Supplier #:  72647-07
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
Catalog # 100489-536
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
3.00 ml, Yes
Price
370,52 $
Log in for availability
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
V-Vials are ideal for: micro mixing, homogenization, centrifugation, concentration, extraction, derivatization, radiosynthesis, hybridization, anaerobic reactions, and catalytic hydrogenation.
  • All components can be autoclaved
  • Graduated and clear
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
Screw cap has solid top and TFE-faced has white (styrene butadiene) rubber liner.
Packaging: They come packed 12 vials in a hinged lid storage box.
Specifications
  • Cap included:
    Yes
  • Capacity:
    3.00 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)