Orders LinkContactUsLinkComponent
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Tough-Spots on laser sheets, round, Orange, 3/8", 3840 labels/pack
  101413-974
 :  77585-O
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
  101413-974
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Labels on Laser Sheets, Round, Electron Microscopy Sciences
Adhesive Tough-Spots on laser sheets, round, Orange, 3/8", 3840 labels/pack
$218.42
Each Pack of 3,840
Tough-Spots® Labels are pre-cut peel-off labels specially for microcentrifuge tube tops. This chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • Polyolefin labels resist most organic solvents and caustic agents
  • They adhere to most plastics, glass and metals without cracking, peeling, or degrading
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Label sheets are top-coated to readily accept laser printing
  • Assortment includes four sheets of blue, green, red, yellow and orange color
Made of polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
  • Color:
    Orange
  • Description:
    Adhesive Tough-Spots on laser sheets, round
  • Packaging:
    3840 labels/pack
  • Diameter:
    3/8"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.