Dulbecco’s Phosphate Buffer Saline – D-PBS, Electron Microscopy Sciences
Product Details & Documents
All of our Dullbecco’s phosphate buffer saline solutions contain: potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose.
- Some formulations may contain additional components
- Store at room temperature
Use the table below as a quick reference for identifying the one that best suits your application. All of the solutions are prepared 1X strength.
Specifications
Specifications
- Concentration:0.19 M
- Pack type:Bottle
- Description:D-PBS, Formula II Phosphate Buffer Saline
- Size:1 l
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



