Orders LinkContactUsLinkComponent
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Square Bottle Series 2, 78.00 ml, Yes
Catalog # 100496-188
Supplier #:  64282
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Catalog # 100496-188
Supplier:  
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Square Bottle Series 2, 78.00 ml, Yes
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A large size storage set. It comes with square clear glass bottles (1 ounce capacity) and black screw caps with plastic linings.
Alpha-numerical indexing system allows quick identification and location of stored items.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Specifications
  • Cap included:
    Yes
  • Capacity:
    78.00 ml
  • Description:
    Square Bottle Series 2
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)