Orders LinkContactUsLinkComponent
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Nickel, 0.7 mil., 50/50 µm, 200/200 µm, 100/100, 250/250 µm
Catalog # 100495-724
Supplier #:  GD1010-NI
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Catalog # 100495-724
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Nickel, 0.7 mil., 50/50 µm, 200/200 µm, 100/100, 250/250 µm
Price
$162.88
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Double grids are used mostly in metallurgical applications for supporting thin metal foils. These grids have a curved securing tab which folds to the curvature of the 'sandwiched' grid.
  • Material: Copper (Cu) or Nickel (Ni)
  • Measures 3.05 mm in diameter
  • Four configurations are available
Specifications
  • Type:
    100/100
  • Material:
    Nickel
  • Thickness:
    0.7 mil.
  • Hole size:
    200/200 µm
  • Bar size:
    50/50 µm
  • Pitch:
    250/250 µm
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)