Orders LinkContactUsLinkComponent
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
5 Thin Film Standards
Catalog # 100495-246
Supplier #:  80080-05
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
Catalog # 100495-246
Supplier:  
CAS Number:  
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
MAC Thin Film Standards for TEM, Electron Microscopy Sciences
5 Thin Film Standards
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These standards are supplied as fine powders that are dispersed onto holey-carbon films and are selected from the range of certified natural and synthetic compounds already in use to prepare bulk analysis standards.
  • The holey-carbon films are supported by a 3.05 mm copper grid (400 mesh)
The Universal set of Thin Film Standards are Ag2Te, BaSO4, Be3Al2Si6O18, Bi2Se3, CaMoO4, CaWO4, CdTe, CeAl2, Cu2S, FeCr2O4, GaAs, Gd3Ga5O12, HgTe, InP, InSb, KAlSi3O8, LaB6, Li2Ta2O6, (Mg,Fe)2SiO4, Na3AlF6, Pb-Ge glass, SrTiO3, TlBr, ZnS, ZrSiO4.
Specifications
Specifications
  • Description:
    5 Thin Film Standards
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.