Orders LinkContactUsLinkComponent
Dropping Bottles, Glass, Electron Microscopy Sciences
Dropping Bottles, Glass, Electron Microscopy Sciences
Amber Dropping Bottle, 50 ml
Catalog # 100490-430
Supplier #:  64221-05
Dropping Bottles, Glass, Electron Microscopy Sciences
Catalog # 100490-430
Supplier:  
Dropping Bottles, Glass, Electron Microscopy Sciences
Amber Dropping Bottle, 50 ml
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.
Specifications
  • Description:
    Amber Dropping Bottle, 50 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)