Orders LinkContactUsLinkComponent
Polyethylene Storage Sheets, Polyview Pages, Electron Microscopy Sciences
Polyethylene Storage Sheets, Polyview Pages, Electron Microscopy Sciences
Film Sleeve Frosted Sealed Flap, 5x7"
Catalog # 100489-898
Supplier #:  73908
Polyethylene Storage Sheets, Polyview Pages, Electron Microscopy Sciences
Catalog # 100489-898
Supplier:  
Polyethylene Storage Sheets, Polyview Pages, Electron Microscopy Sciences
Polyethylene Storage Sheets, Polyview Pages, Electron Microscopy Sciences
Film Sleeve Frosted Sealed Flap, 5x7"
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sheets are completely transparent. Fits all U.S. standard 3-ring binders: 8 1/4 x 1 1/4". Convenient. Offers good protection for your valuable negatives and prints.
Specifications
  • Description:
    Film Sleeve Frosted Sealed Flap
  • Size:
    5x7"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)