Orders LinkContactUsLinkComponent
Cotton Tipped Applicators, Electron Microscopy Sciences
Handy Vial Pack, Sterile, 10 Vials/Pack
  100496-340
Cotton Tipped Applicators, Electron Microscopy Sciences
  100496-340
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Handy Vial Pack, Sterile, 10 Vials/Pack
$94.20
Each
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.
  • Sterility:
    Sterile
  • Description:
    Handy Vial Pack
  • Packaging:
    10 Vials/Pack
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.