Orders LinkContactUsLinkComponent
Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
Cryo labels, 25×50 mm, 10 packs/lot
Catalog # 100496-126
Supplier #:  62047
Cryo Labels, Electron Microscopy Sciences
Catalog # 100496-126
Supplier:  
Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
Cryo labels, 25×50 mm, 10 packs/lot
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen.
  • Will not peel or shrink
They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. Ten sheets of labels: five preprinted, five plain.
 : Packaged as 20 labels/sheet.
Specifications
  • Description:
    Cryo labels
  • Dimensions:
    25×50 mm
  • Packaging:
    10 packs/lot
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.