Tissue Chopper, Electron Microscopy Sciences
Product Details & Documents
The TC-1 Tissue Chopper is designed to facilitate the cutting of 100 to 1000 micron thick brain tissue slices for use in recording experiments.
The chopper includes a guillotine arm with adjustments to assure blade alignment. The brain sample is moved in respect to the arm with a stage whose position can be read with 2 to 3 micron precision on a digital micrometre scale. The stage is made with a removable "PTFE" surface plate to facilitate cleaning.
Specifications
- Description:Replacement PFTE Plate
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.