Orders LinkContactUsLinkComponent
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
Capinsert Assorted Colors
Catalog # 101454-056
Supplier #:  61806-AS
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
Catalog # 101454-056
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
Capinsert Assorted Colors
Price
$15.74
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tamperproof multi purpose container for mailing, staining or storing microscope slides. Constructed of extra strong and opaque clear polypropylene, and high density polyethylene leakproof tamper evident screw cap (The LockMailer can also be used without the tamper evident feature by not used the lock tab), it will hold up to 4 standard 3 x 1” or 25 x 75mm slides vertically.
Inside chambers are slotted to keep slides safely separated. For color coding purposes, use Capinsert™ (#61805-series) that may be inserted on top of closure. This LockMailer™ can be used as small staining vessel. This unit is designed for maximum stability on a bench top with liquid volume only 14 ml. Each standard volume of one slide is 2 ml, therefore when you want to stain 4 slides in this vessel you need only 6 ml of staining solution. Color coded CapInsert™ is available for the LockMailer™. Measures 35 x87 mm H (1 3/8” x 3 7/16”H). Packaging: 100 per pack. 
Specifications
  • Description:
    Capinsert Assorted Colors
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)